Product Details

SNP ID
rs217154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31517900 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCAGAATAAGAGGTCTCCACTGT[G/T]CTGAGTTATGGGAGAAAGATGGGTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC129 PubMed Links

Gene Details

Gene
CCDC129
Gene Name
coiled-coil domain containing 129
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257967.1 Intron NP_001244896.1
NM_001257968.1 Intron NP_001244897.1
NM_194300.3 Intron NP_919276.2
XM_011515213.1 Intron XP_011513515.1
XM_017011871.1 Intron XP_016867360.1
XM_017011872.1 Intron XP_016867361.1

View Full Product Details