Product Details
- SNP ID
-
rs193280665
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:13107002 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCAAAGTGACAGTGCCTTTTGTC[C/T]GTTTAAGGATGGCAACAGCTTCTTC
- Phenotype
-
MIM: 603785
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MPDZ
PubMed Links
Gene Details
- Gene
- MPDZ
- Gene Name
- multiple PDZ domain crumbs cell polarity complex component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001261406.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q2026R |
NP_001248335.1 |
NM_001261407.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1997R |
NP_001248336.1 |
NM_003829.4 |
6311 |
Missense Mutation |
CAG,CGG |
Q2030R |
NP_003820.2 |
XM_005251622.4 |
6311 |
Missense Mutation |
CAG,CGG |
Q2059R |
XP_005251679.1 |
XM_005251623.4 |
6311 |
Missense Mutation |
CAG,CGG |
Q1989R |
XP_005251680.1 |
XM_006716885.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q2059R |
XP_006716948.1 |
XM_006716886.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q2059R |
XP_006716949.1 |
XM_006716887.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q2059R |
XP_006716950.1 |
XM_006716888.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q2030R |
XP_006716951.1 |
XM_006716889.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q2026R |
XP_006716952.1 |
XM_006716891.3 |
6311 |
Missense Mutation |
CAG,CGG |
Q1997R |
XP_006716954.1 |
XM_017015252.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q2031R |
XP_016870741.1 |
XM_017015253.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1997R |
XP_016870742.1 |
XM_017015254.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1989R |
XP_016870743.1 |
XM_017015255.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1989R |
XP_016870744.1 |
XM_017015256.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1960R |
XP_016870745.1 |
XM_017015257.1 |
6311 |
Missense Mutation |
CAG,CGG |
Q1960R |
XP_016870746.1 |
XM_017015258.1 |
6311 |
Intron |
|
|
XP_016870747.1 |
XM_017015259.1 |
6311 |
Intron |
|
|
XP_016870748.1 |
View Full Product Details