Product Details

SNP ID
rs193272374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8319287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGTAGGAGACTGTTTTGGGCT[C/T]AGGGGCTTTGCAGATCTGTGGAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGK223 PubMed Links

Gene Details

Gene
SGK223
Gene Name
homolog of rat pragma of Rnd2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080826.2 3763 Missense Mutation AAG,GAG K1030E NP_001074295.2
XM_005272369.4 3763 Missense Mutation AAG,GAG K1030E XP_005272426.2
XM_005272370.4 3763 Missense Mutation AAG,GAG K1030E XP_005272427.2
XM_011543813.2 3763 Intron XP_011542115.1

View Full Product Details