Product Details

SNP ID
rs193271462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50191061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCAAACTTCATGACCTCTTGAA[G/T]AAGACCTCCTACACTGTACCCCTTC
Phenotype
MIM: 300185
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP4 PubMed Links

Gene Details

Gene
AKAP4
Gene Name
A-kinase anchoring protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003886.2 2588 Missense Mutation ATT,CTT I822L NP_003877.2
NM_139289.1 2588 Missense Mutation ATT,CTT I813L NP_647450.1

View Full Product Details