Product Details
- SNP ID
-
rs311680
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:73424476 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTATGTATTTCCTGGCCAGGCACGG[T/C]GGAGAGCTGTTTTTCAGTAAGAACT
- Phenotype
-
MIM: 606286
MIM: 613973
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX43
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7754197] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX43
- Gene Name
- DEAD-box helicase 43
There are no transcripts associated with this gene.
- Gene
- MB21D1
- Gene Name
- Mab-21 domain containing 1
View Full Product Details