Product Details

SNP ID
rs311806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11485113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCCCCAAACTTCTGACCTCCTG[C/T]GATATCAGAAGCTCTGGCCCTCAGG
Phenotype
MIM: 603458 MIM: 611371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELAVL3 PubMed Links

Gene Details

Gene
ELAVL3
Gene Name
ELAV like neuron-specific RNA binding protein 3
There are no transcripts associated with this gene.

Gene
ZNF653
Gene Name
zinc finger protein 653
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138783.3 Intron NP_620138.2

View Full Product Details