Product Details

SNP ID
rs313536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25254325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGGGAAAGTAAACAGCAAACA[C/T]GTAATGTGGATATTTAATAGATAGT
Phenotype
MIM: 612101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI4K2B PubMed Links

Gene Details

Gene
PI4K2B
Gene Name
phosphatidylinositol 4-kinase type 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018323.3 Intron NP_060793.2
XM_005248174.1 Intron XP_005248231.1
XM_005248175.4 Intron XP_005248232.1

View Full Product Details