Product Details
- SNP ID
-
rs385101
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2885038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCGGGGCTGGAGCCAGGACCGGG[A/G]CTGGGGGCGGGGTGGACGCCGGGGC
- Phenotype
-
MIM: 600856
MIM: 603240
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDKN1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs565544512] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDKN1C
- Gene Name
- cyclin dependent kinase inhibitor 1C
- Gene
- SLC22A18AS
- Gene Name
- solute carrier family 22 member 18 antisense
There are no transcripts associated with this gene.
View Full Product Details