Product Details

SNP ID
rs5745299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10680163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAATGTGGTGTCTCATGTCCCT[A/G]GGGAGACAAAGGGCAGGTTTCTTAT
Phenotype
MIM: 600954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAP PubMed Links

Gene Details

Gene
DAP
Gene Name
death associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291963.1 1366 UTR 3 NP_001278892.1
NM_004394.2 1366 UTR 3 NP_004385.1

View Full Product Details