Product Details

SNP ID
rs5851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11721468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATGATTTACACAGTATACTGGA[G/A]GATATGCATAGGTTACATGCAAATA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 2129 UTR 3 NP_001073962.1
NM_001297610.1 2129 UTR 3 NP_001284539.1
NM_017507.1 2129 UTR 3 NP_059977.1
XM_017026941.1 2129 UTR 3 XP_016882430.1
XM_017026942.1 2129 UTR 3 XP_016882431.1

View Full Product Details