Product Details
- SNP ID
-
rs6001054
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38218088 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTTCCAGCACTTTCACATTGAG[A/G]TCTGATTTTCTACAGCCAAGTGGAA
- Phenotype
-
MIM: 604877
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAFF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6001053] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAFF
- Gene Name
- MAF bZIP transcription factor F
There are no transcripts associated with this gene.
- Gene
- SNORA92
- Gene Name
- small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.
- Gene
- TMEM184B
- Gene Name
- transmembrane protein 184B
View Full Product Details