Product Details
- SNP ID
-
rs6003367
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:22888116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGGAGCTGGGCCCTGGTCCCAG[C/G]CAGCGCTGGCCCCTGCTGCTGCTGG
- Phenotype
-
MIM: 147220
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IGLC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs382768] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGLC1
- Gene Name
- immunoglobulin lambda constant 1
There are no transcripts associated with this gene.
- Gene
- IGLL5
- Gene Name
- immunoglobulin lambda like polypeptide 5
- Gene
- MIR5571
- Gene Name
- microRNA 5571
There are no transcripts associated with this gene.
View Full Product Details