Product Details

SNP ID
rs6008357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46256207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCTCCACCCTGGCCAGGTCCC[G/T]CTCTCAAATCTGGTTCTCAAAACCA
Phenotype
MIM: 604670
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDPF1 PubMed Links

Gene Details

Gene
CDPF1
Gene Name
cysteine rich DPF motif domain containing 1
There are no transcripts associated with this gene.

Gene
PKDREJ
Gene Name
polycystin (PKD) family receptor for egg jelly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006071.1 7116 UTR 3 NP_006062.1

View Full Product Details