Product Details

SNP ID
rs6013414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37905969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTAGCAGATTGGGGCCTGATC[A/G]TTTCATTATTCCTGGGCTCCGTGTG
Phenotype
MIM: 616537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VSTM2L PubMed Links

Gene Details

Gene
VSTM2L
Gene Name
V-set and transmembrane domain containing 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080607.2 Intron NP_542174.1
XM_011528530.1 Intron XP_011526832.1

View Full Product Details