Product Details

SNP ID
rs6042671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:14324823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGGAAGCACATTTACATTACC[A/G]TTTTTGTGAAGTCCTTTATTTTCAA
Phenotype
MIM: 604808 MIM: 611567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLRT3 PubMed Links

Gene Details

Gene
FLRT3
Gene Name
fibronectin leucine rich transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013281.3 3849 UTR 3 NP_037413.1
NM_198391.2 3849 UTR 3 NP_938205.1
XM_005260682.4 3849 UTR 3 XP_005260739.1
XM_011529204.2 3849 UTR 3 XP_011527506.1
XM_011529205.2 3849 UTR 3 XP_011527507.1
Gene
MACROD2
Gene Name
MACRO domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033087.1 3849 Intron NP_001028259.1
NM_080676.5 3849 Intron NP_542407.2
XM_017027675.1 3849 Intron XP_016883164.1
XM_017027676.1 3849 Intron XP_016883165.1
XM_017027677.1 3849 Intron XP_016883166.1

View Full Product Details