Product Details

SNP ID
rs6019944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49633767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAGATCGCCTGACAACACTGCC[A/G]CATACACATACGGCACCCAGGACTC
Phenotype
MIM: 604016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALT5 PubMed Links

Gene Details

Gene
B4GALT5
Gene Name
beta-1,4-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004776.3 3906 UTR 3 NP_004767.1

View Full Product Details