Product Details

SNP ID
rs612211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55639105 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGACACTCAGGTTGGATAATTGTC[T/C]AAGTAGACCAGGGGTTCTCAGCTAG
Phenotype
MIM: 613478
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CCDC106 PubMed Links

Gene Details

Gene
CCDC106
Gene Name
coiled-coil domain containing 106
There are no transcripts associated with this gene.

Gene
LOC107983998
Gene Name
uncharacterized LOC107983998
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527018.2 Intron XP_011525320.1
XM_011527021.1 Intron XP_011525323.1
Gene
ZNF580
Gene Name
zinc finger protein 580
There are no transcripts associated with this gene.

Gene
ZNF581
Gene Name
zinc finger protein 581
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016535.3 Intron NP_057619.1
XM_006723240.3 Intron XP_006723303.1
XM_017026867.1 Intron XP_016882356.1

View Full Product Details