Product Details

SNP ID
rs6130531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44113049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTCAACACCCACCCCACCCA[A/G]CTTCCCTGGGAGGCCTCAGCCCACA
Phenotype
MIM: 605267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JPH2 PubMed Links

Gene Details

Gene
JPH2
Gene Name
junctophilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020433.4 3433 UTR 3 NP_065166.2
NM_175913.3 3433 Intron NP_787109.2
XM_006723833.3 3433 Intron XP_006723896.1

View Full Product Details