Product Details

SNP ID
rs6720068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37168495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAAAACCTAATGTGATTTCTGTT[A/G]TAAAATTAACAATATCCATTGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT6B1 PubMed Links

Gene Details

Gene
SULT6B1
Gene Name
sulfotransferase family 6B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032377.1 Intron NP_001027549.1

View Full Product Details