Product Details

SNP ID
hCV189162553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:1292791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGGCGTCCTCAAGAGTCCACA[A/G]AGAAAAGACGCAAGACAAAGTCAGA
Phenotype
MIM: 601328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
There are no transcripts associated with this gene.

Gene
MIR6726
Gene Name
microRNA 6726
There are no transcripts associated with this gene.

Gene
SCNN1D
Gene Name
sodium channel epithelial 1 delta subunit
There are no transcripts associated with this gene.

View Full Product Details