Product Details
- SNP ID
-
rs6664239
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:11740464 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTATCAAACCAAAGTGACGTCA[C/T]GCAGAGGAGACTGTGTGTGTGCTAA
- Phenotype
-
MIM: 608729
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGTRAP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11121816] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGTRAP
- Gene Name
- angiotensin II receptor associated protein
View Full Product Details