Product Details

SNP ID
rs650102
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:23381199 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CAGAGTTTGCAGATTACAGCCATAA[A/G]CCACCGTGCCCAGCCCTCTGCAGTT
Phenotype
MIM: 604128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEA3 PubMed Links

Gene Details

Gene
TCEA3
Gene Name
transcription elongation factor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003196.2 1460 UTR 3 NP_003187.1
XM_006710864.1 1460 Intron XP_006710927.1
XM_011542053.2 1460 UTR 3 XP_011540355.1
XM_017002200.1 1460 Intron XP_016857689.1
XM_017002201.1 1460 Intron XP_016857690.1
XM_017002202.1 1460 Intron XP_016857691.1
XM_017002203.1 1460 Intron XP_016857692.1
XM_017002204.1 1460 Intron XP_016857693.1
Gene
ZNF436-AS1
Gene Name
ZNF436 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details