Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015320.3 | 2111 | Intron | NP_056135.2 | ||
NM_032995.2 | 2111 | Intron | NP_127462.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001009993.3 | 2111 | UTR 3 | NP_001009993.2 | ||
NM_001321743.1 | 2111 | UTR 3 | NP_001308672.1 | ||
NM_001321744.1 | 2111 | UTR 3 | NP_001308673.1 | ||
NM_001321745.1 | 2111 | UTR 3 | NP_001308674.1 | ||
NM_001321746.1 | 2111 | UTR 3 | NP_001308675.1 | ||
NM_001321747.1 | 2111 | UTR 3 | NP_001308676.1 | ||
NM_001321748.1 | 2111 | UTR 3 | NP_001308677.1 | ||
XM_011510594.2 | 2111 | UTR 3 | XP_011508896.1 | ||
XM_017003327.1 | 2111 | UTR 3 | XP_016858816.1 | ||
XM_017003328.1 | 2111 | UTR 3 | XP_016858817.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001277406.1 | 2111 | Intron | NP_001264335.1 | ||
XM_011511670.2 | 2111 | Intron | XP_011509972.1 | ||
XM_017004732.1 | 2111 | Intron | XP_016860221.1 | ||
XM_017004733.1 | 2111 | Intron | XP_016860222.1 | ||
XM_017004734.1 | 2111 | Intron | XP_016860223.1 | ||
XM_017004735.1 | 2111 | Intron | XP_016860224.1 |