Product Details

SNP ID
rs6790275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183643608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGAATTTAAACCATATTCCTTT[C/T]TTTAACCAGGGATCACTTAAGCCAA
Phenotype
MIM: 611295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL24 PubMed Links

Gene Details

Gene
KLHL24
Gene Name
kelch like family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017644.3 Intron NP_060114.2
XM_005247552.2 Intron XP_005247609.1
XM_005247553.1 Intron XP_005247610.1
XM_005247554.2 Intron XP_005247611.1
XM_005247555.1 Intron XP_005247612.1
XM_005247556.1 Intron XP_005247613.1
XM_011512938.2 Intron XP_011511240.1
XM_017006650.1 Intron XP_016862139.1
XM_017006651.1 Intron XP_016862140.1
XM_017006652.1 Intron XP_016862141.1
XM_017006653.1 Intron XP_016862142.1
XM_017006654.1 Intron XP_016862143.1
XM_017006655.1 Intron XP_016862144.1
XM_017006656.1 Intron XP_016862145.1
XM_017006657.1 Intron XP_016862146.1
XM_017006658.1 Intron XP_016862147.1
XM_017006659.1 Intron XP_016862148.1
XM_017006660.1 Intron XP_016862149.1
XM_017006661.1 Intron XP_016862150.1
XM_017006662.1 Intron XP_016862151.1
XM_017006663.1 Intron XP_016862152.1

View Full Product Details