Product Details

SNP ID
rs7151871
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64508738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGGGAAAATGCTTTAAATTTTAA[C/T]TTTAATTTTAATTTCCAACCACTAT
Phenotype
MIM: 616578 MIM: 194541
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC102723809 PubMed Links
Additional Information
For this assay, SNP(s) [rs201815597] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
ZBTB1
Gene Name
zinc finger and BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123329.1 Intron NP_001116801.1
NM_014950.2 Intron NP_055765.2
XM_005267410.1 Intron XP_005267467.1
XM_011536566.2 Intron XP_011534868.1
XM_011536568.2 Intron XP_011534870.1
XM_017021094.1 Intron XP_016876583.1
XM_017021095.1 Intron XP_016876584.1
Gene
ZBTB25
Gene Name
zinc finger and BTB domain containing 25
There are no transcripts associated with this gene.

View Full Product Details