Product Details

SNP ID
rs7189908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:189108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAACAATAGAAATTTTAAACTA[C/T]AAAAGATGAGTTGTATTCAGCAAAT
Phenotype
MIM: 142240 MIM: 607782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HBQ1 PubMed Links

Gene Details

Gene
HBQ1
Gene Name
hemoglobin subunit theta 1
There are no transcripts associated with this gene.

Gene
LUC7L
Gene Name
LUC7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320226.1 2476 UTR 3 NP_001307155.1
NM_018032.4 2476 UTR 3 NP_060502.1
NM_201412.2 2476 UTR 3 NP_958815.1
XM_005255427.3 2476 UTR 3 XP_005255484.1
XM_005255429.3 2476 UTR 3 XP_005255486.1
XM_011522561.2 2476 UTR 3 XP_011520863.1
XM_017023437.1 2476 UTR 3 XP_016878926.1
XM_017023438.1 2476 UTR 3 XP_016878927.1
XM_017023439.1 2476 UTR 3 XP_016878928.1
XM_017023440.1 2476 UTR 3 XP_016878929.1

View Full Product Details