Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001251888.1 | Intron | NP_001238817.1 | |||
NM_024083.3 | Intron | NP_076988.1 | |||
XM_011523601.2 | Intron | XP_011521903.1 | |||
XM_011523602.2 | Intron | XP_011521904.1 | |||
XM_017025036.1 | Intron | XP_016880525.1 | |||
XM_017025037.1 | Intron | XP_016880526.1 | |||
XM_017025038.1 | Intron | XP_016880527.1 | |||
XM_017025039.1 | Intron | XP_016880528.1 | |||
XM_017025040.1 | Intron | XP_016880529.1 | |||
XM_017025041.1 | Intron | XP_016880530.1 | |||
XM_017025042.1 | Intron | XP_016880531.1 | |||
XM_017025043.1 | Intron | XP_016880532.1 | |||
XM_017025044.1 | Intron | XP_016880533.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271006.1 | Intron | NP_001257935.1 | |||
NM_001271007.1 | Intron | NP_001257936.1 | |||
NM_144998.3 | Intron | NP_659435.2 | |||
XM_005256339.2 | Intron | XP_005256396.1 | |||
XM_017024326.1 | Intron | XP_016879815.1 | |||
XM_017024327.1 | Intron | XP_016879816.1 | |||
XM_017024328.1 | Intron | XP_016879817.1 | |||
XM_017024329.1 | Intron | XP_016879818.1 |