Product Details
- SNP ID
-
rs7205390
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:68103570 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTAATTTTACCAAGTTGTATTA[A/C]TTGAGTTCTTTTGTTGCTTGTGCTT
- Phenotype
-
MIM: 602698
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NFATC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77996485] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NFATC3
- Gene Name
- nuclear factor of activated T-cells 3
View Full Product Details