Product Details

SNP ID
rs7216595
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82021779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGGCCTTTTGGCTACTTTAAC[C/T]TGGTGTTCTCACAACCACCCAGTGA
Phenotype
MIM: 606236 MIM: 602050 MIM: 615128
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs4077074] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
There are no transcripts associated with this gene.

Gene
LRRC45
Gene Name
leucine rich repeat containing 45
There are no transcripts associated with this gene.

Gene
RAC3
Gene Name
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
There are no transcripts associated with this gene.

Gene
STRA13
Gene Name
stimulated by retinoic acid 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271006.1 Intron NP_001257935.1
NM_001271007.1 Intron NP_001257936.1
NM_144998.3 Intron NP_659435.2
XM_005256339.2 Intron XP_005256396.1
XM_017024326.1 Intron XP_016879815.1
XM_017024327.1 Intron XP_016879816.1
XM_017024328.1 Intron XP_016879817.1
XM_017024329.1 Intron XP_016879818.1

View Full Product Details