Product Details

SNP ID
rs7214211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32329031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATATTCAGTTCCATGACACCA[C/T]TTTCCTTGTCTACAGTAGTGATACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 4011 UTR 3 NP_071739.2
XM_005258022.3 4011 UTR 3 XP_005258079.1
XM_017024940.1 4011 UTR 3 XP_016880429.1
XM_017024941.1 4011 UTR 3 XP_016880430.1
Gene
RHBDL3
Gene Name
rhomboid like 3
There are no transcripts associated with this gene.

View Full Product Details