Product Details

SNP ID
rs72475636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69701257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGAGAAATTAGTATCACTTTCT[C/T]CATACAGAAAACGAAATGACTTCAA
Phenotype
MIM: 603891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF19 PubMed Links

Gene Details

Gene
FGF19
Gene Name
fibroblast growth factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005117.2 Intron NP_005108.1

View Full Product Details