Product Details
- SNP ID
-
rs7293280
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:8846717 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAAACCTCTCACCCCTTACCCCA[C/T]CAGGCATTGAACCATACACGGAGCT
- Phenotype
-
MIM: 142992
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HMX1
PubMed Links
Gene Details
- Gene
- HMX1
- Gene Name
- H6 family homeobox 1
View Full Product Details