Product Details

SNP ID
rs7258210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15834804 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGCCCAGCCAAGGCCATGCTTCC[A/T]TGCAACACTCCTTCCAAATGGCCAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UCA1 PubMed Links

Gene Details

Gene
UCA1
Gene Name
urothelial cancer associated 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details