Product Details

SNP ID
rs733699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39503873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCCTAGGTGGGCTGGTCACCTC[G/A]TCTCTGTGGGCCTCAGTTTCCCTAT
Phenotype
MIM: 602768
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016941.3 Intron NP_058637.1
NM_203486.2 Intron NP_982353.1

View Full Product Details