Product Details

SNP ID
rs7316528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31654395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGCAGTTAATCATACACCTAC[C/T]ATATGATCTAGCCATTGCACTCCTA
Phenotype
MIM: 615256
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETFBKMT PubMed Links

Gene Details

Gene
ETFBKMT
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135863.1 Intron NP_001129335.1
NM_001135864.1 Intron NP_001129336.1
NM_173802.3 Intron NP_776163.1

View Full Product Details