Product Details

SNP ID
rs7363950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46023753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCACTGCACCCTTAGTGCACA[C/T]GTCCCGGTGGATGGCAGTCGTTTTC
Phenotype
MIM: 120361 MIM: 606726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP9 PubMed Links

Gene Details

Gene
MMP9
Gene Name
matrix metallopeptidase 9
There are no transcripts associated with this gene.

Gene
SLC12A5
Gene Name
solute carrier family 12 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134771.1 Intron NP_001128243.1
NM_020708.4 Intron NP_065759.1
XM_017027981.1 Intron XP_016883470.1

View Full Product Details