Product Details

SNP ID
rs7590385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241703966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGGCAGCGTGGCTACACTGCTC[A/G]ATGCACTGGATTTCTGGGTAAAGTA
Phenotype
MIM: 608525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ING5 PubMed Links

Gene Details

Gene
ING5
Gene Name
inhibitor of growth family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005247047.2 Intron XP_005247104.1
XM_017005097.1 Intron XP_016860586.1
XM_017005098.1 Intron XP_016860587.1
XM_017005099.1 Intron XP_016860588.1
XM_017005100.1 Intron XP_016860589.1
XM_017005101.1 Intron XP_016860590.1
XM_017005102.1 Intron XP_016860591.1

View Full Product Details