Product Details

SNP ID
rs7607995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74424186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTATAGTTAATTAAGACTTTAT[A/C]ATACTGATGTGAAATGAGGAGTTGG
Phenotype
MIM: 602288
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
There are no transcripts associated with this gene.

Gene
RTKN
Gene Name
rhotekin
There are no transcripts associated with this gene.

Gene
WDR54
Gene Name
WD repeat domain 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320823.1 Intron NP_001307752.1
NM_001320824.1 Intron NP_001307753.1
NM_001320825.1 Intron NP_001307754.1
NM_032118.3 Intron NP_115494.1
XM_017005064.1 Intron XP_016860553.1

View Full Product Details