Product Details

SNP ID
rs7570571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:953677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAAAATCCATGCATTACAAAG[C/T]AGGCAGTTACCTGACTTTACCAGGA
Phenotype
MIM: 608715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101060391 PubMed Links

Gene Details

Gene
LOC101060391
Gene Name
uncharacterized LOC101060391
There are no transcripts associated with this gene.

Gene
SNTG2
Gene Name
syntrophin gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018968.3 Intron NP_061841.2
XM_011510366.2 Intron XP_011508668.1
XM_017004361.1 Intron XP_016859850.1
XM_017004362.1 Intron XP_016859851.1
XM_017004363.1 Intron XP_016859852.1
XM_017004364.1 Intron XP_016859853.1
XM_017004365.1 Intron XP_016859854.1
XM_017004366.1 Intron XP_016859855.1
XM_017004367.1 Intron XP_016859856.1
XM_017004368.1 Intron XP_016859857.1
XM_017004369.1 Intron XP_016859858.1
XM_017004370.1 Intron XP_016859859.1
XM_017004371.1 Intron XP_016859860.1
XM_017004372.1 Intron XP_016859861.1
XM_017004373.1 Intron XP_016859862.1
XM_017004374.1 Intron XP_016859863.1

View Full Product Details