Product Details

SNP ID
rs757873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31134971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCAGCTAGGTGTCGCAGGGGGGA[G/A]TACTTATTAAGTGCTAAGCACTGTA
Phenotype
MIM: 606481 MIM: 611651
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
INPP5J PubMed Links

Gene Details

Gene
INPP5J
Gene Name
inositol polyphosphate-5-phosphatase J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002837.2 2535 Intron NP_001002837.1
NM_001284285.1 2535 Intron NP_001271214.1
NM_001284286.1 2535 Intron NP_001271215.1
NM_001284287.1 2535 Intron NP_001271216.1
NM_001284288.1 2535 Intron NP_001271217.1
NM_001284289.1 2535 Intron NP_001271218.1
XM_011530143.1 2535 Intron XP_011528445.1
XM_017028771.1 2535 Intron XP_016884260.1
XM_017028772.1 2535 Intron XP_016884261.1
XM_017028773.1 2535 Intron XP_016884262.1
Gene
PLA2G3
Gene Name
phospholipase A2 group III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015715.4 2535 UTR 3 NP_056530.2
XM_011530204.1 2535 UTR 3 XP_011528506.1
XM_011530205.1 2535 UTR 3 XP_011528507.1

View Full Product Details