Product Details

SNP ID
rs7578864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27099582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTTCAGCCTCTCCCAGGGTGA[C/G]TATGGCAGCAGGAGGGGAAAAGGAC
Phenotype
MIM: 606137 MIM: 614058
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CGREF1 PubMed Links

Gene Details

Gene
CGREF1
Gene Name
cell growth regulator with EF-hand domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166239.1 1032 Intron NP_001159711.1
NM_001166240.1 1032 UTR 3 NP_001159712.1
NM_001301324.1 1032 UTR 3 NP_001288253.1
NM_006569.5 1032 Intron NP_006560.3
Gene
KHK
Gene Name
ketohexokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000221.2 1032 Intron NP_000212.1
NM_006488.2 1032 Intron NP_006479.1
XM_005264294.3 1032 Intron XP_005264351.1
XM_005264296.3 1032 Intron XP_005264353.1
XM_005264298.3 1032 Intron XP_005264355.1
XM_006712008.3 1032 Intron XP_006712071.1
XM_006712009.3 1032 Intron XP_006712072.1
XM_006712010.3 1032 Intron XP_006712073.1
XM_006712011.3 1032 Intron XP_006712074.1
XM_006712012.3 1032 Intron XP_006712075.1
XM_006712013.3 1032 Intron XP_006712076.1
XM_006712014.3 1032 Intron XP_006712077.1
XM_017004060.1 1032 Intron XP_016859549.1
XM_017004061.1 1032 Intron XP_016859550.1

View Full Product Details