Product Details

SNP ID
rs7585597
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238173399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCACCAGAGCAGAAACCACATT[C/T]TGTTTATCTCTGCACCCCCAGGGCC
Phenotype
MIM: 615099
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ERFE PubMed Links

Gene Details

Gene
ERFE
Gene Name
erythroferrone
There are no transcripts associated with this gene.

Gene
ILKAP
Gene Name
ILK associated serine/threonine phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030768.2 Intron NP_110395.1
XM_005246106.1 Intron XP_005246163.1
XM_006712784.1 Intron XP_006712847.1
XM_011511946.2 Intron XP_011510248.1
XM_017005056.1 Intron XP_016860545.1
XM_017005057.1 Intron XP_016860546.1
XM_017005058.1 Intron XP_016860547.1

View Full Product Details