Product Details

SNP ID
rs7682250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44178238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTAAAAGGATCTAGTATAATGA[C/T]TCTCAACAATGAGAGTATATCAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD8 PubMed Links

Gene Details

Gene
KCTD8
Gene Name
potassium channel tetramerization domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198353.2 Intron NP_938167.1
XM_011513690.2 Intron XP_011511992.1

View Full Product Details