Product Details

SNP ID
rs768531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21027974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGAACTGAATAGTAGCTTCCCAT[A/G]TATTATCCCCTAATACCGTAAGGTT
Phenotype
MIM: 605272 MIM: 616956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6717 PubMed Links

Gene Details

Gene
MIR6717
Gene Name
microRNA 6717
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 Intron NP_001269140.1
NM_001282212.1 Intron NP_001269141.1
NM_001282213.1 Intron NP_001269142.1
NM_001282214.1 Intron NP_001269143.1
NM_001282215.1 Intron NP_001269144.1
NM_001282216.1 Intron NP_001269145.1
NM_001320329.1 Intron NP_001307258.1
NM_016250.2 Intron NP_057334.1
NM_201535.1 Intron NP_963293.1
NM_201536.1 Intron NP_963294.1
NM_201537.1 Intron NP_963831.1
NM_201538.1 Intron NP_963832.1
NM_201539.1 Intron NP_963833.1
NM_201540.1 Intron NP_963834.1
NM_201541.1 Intron NP_963835.1
XM_011536996.2 Intron XP_011535298.1
XM_011536997.1 Intron XP_011535299.1
XM_011536998.1 Intron XP_011535300.1
XM_011536999.1 Intron XP_011535301.1
XM_011537001.1 Intron XP_011535303.1
XM_011537002.1 Intron XP_011535304.1
XM_017021480.1 Intron XP_016876969.1
XM_017021481.1 Intron XP_016876970.1
XM_017021482.1 Intron XP_016876971.1
XM_017021483.1 Intron XP_016876972.1
XM_017021484.1 Intron XP_016876973.1
XM_017021485.1 Intron XP_016876974.1
XM_017021486.1 Intron XP_016876975.1
XM_017021487.1 Intron XP_016876976.1
XM_017021488.1 Intron XP_016876977.1
XM_017021489.1 Intron XP_016876978.1
XM_017021490.1 Intron XP_016876979.1
XM_017021491.1 Intron XP_016876980.1
XM_017021492.1 Intron XP_016876981.1
XM_017021493.1 Intron XP_016876982.1
XM_017021494.1 Intron XP_016876983.1
XM_017021495.1 Intron XP_016876984.1
XM_017021496.1 Intron XP_016876985.1
XM_017021497.1 Intron XP_016876986.1
Gene
RNASE13
Gene Name
ribonuclease A family member 13 (inactive)
There are no transcripts associated with this gene.

Gene
TPPP2
Gene Name
tubulin polymerization promoting protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173846.4 Intron NP_776245.2
XM_005267324.4 Intron XP_005267381.1
XM_011536416.1 Intron XP_011534718.1
XM_011536417.2 Intron XP_011534719.1
XM_011536418.1 Intron XP_011534720.1
XM_011536420.2 Intron XP_011534722.1
XM_017020966.1 Intron XP_016876455.1

View Full Product Details