Product Details

SNP ID
rs7700841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134854895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATATATTTGTCTTTTATTTTTGG[C/T]AGAAAATGATGCATCCTGTTGCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf24 PubMed Links

Gene Details

Gene
C5orf24
Gene Name
chromosome 5 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135586.1 Intron NP_001129058.1
NM_001300894.1 Intron NP_001287823.1
NM_152409.3 Intron NP_689622.2
XM_005271889.3 Intron XP_005271946.1
XM_017009049.1 Intron XP_016864538.1
XM_017009050.1 Intron XP_016864539.1

View Full Product Details