Product Details

SNP ID
rs7767
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35432172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTGGCTGTCCTAGTAGTGGACG[C/T]GGCATCAGCCTACCAGCAATGGAGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PAMR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs1048193] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PAMR1
Gene Name
peptidase domain containing associated with muscle regeneration 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001991.2 2844 UTR 3 NP_001001991.1
NM_001282675.1 2844 UTR 3 NP_001269604.1
NM_001282676.1 2844 UTR 3 NP_001269605.1
NM_015430.3 2844 UTR 3 NP_056245.2

View Full Product Details