Product Details

SNP ID
rs7908974
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5639427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCAAGTTGGCCAGGCTGGTTTC[A/G]AACTCCTGACCTCAGATGCCCCAGA
Phenotype
MIM: 615055
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ASB13 PubMed Links

Gene Details

Gene
ASB13
Gene Name
ankyrin repeat and SOCS box containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024701.3 2157 UTR 3 NP_078977.2

View Full Product Details