Product Details

SNP ID
rs7939655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128894489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCATTGAGGGCTGGCAGTGTT[A/T]CCAAGTGCGGATCTGTCATCAGATA
Phenotype
MIM: 600734
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf45 PubMed Links

Gene Details

Gene
C11orf45
Gene Name
chromosome 11 open reading frame 45
There are no transcripts associated with this gene.

Gene
KCNJ5
Gene Name
potassium voltage-gated channel subfamily J member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000890.3 Intron NP_000881.3
XM_011542809.2 Intron XP_011541111.1
XM_011542810.2 Intron XP_011541112.1

View Full Product Details