Product Details
- SNP ID
-
rs7942794
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18084522 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATCTCACTGTTCCTCCTCAAAGA[C/T]ACCAGGCATACTTCTGGTTGAAGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SAAL1
PubMed Links
Gene Details
- Gene
- SAAL1
- Gene Name
- serum amyloid A like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138421.2 |
|
Intron |
|
|
NP_612430.2 |
View Full Product Details