Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308396.1 | Intron | NP_001295325.1 | |||
NM_032112.2 | Intron | NP_115488.2 | |||
NM_176792.2 | Intron | NP_789762.1 | |||
NM_176793.1 | Intron | NP_789763.1 | |||
NM_176794.1 | Intron | NP_789764.1 | |||
XM_005270231.2 | Intron | XP_005270288.1 | |||
XM_006718035.3 | Intron | XP_006718098.1 | |||
XM_017016790.1 | Intron | XP_016872279.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001203244.1 | Intron | NP_001190173.1 | |||
NM_017893.3 | Intron | NP_060363.2 | |||
XM_005270008.2 | Intron | XP_005270065.1 |